Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F2
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
Brand: Fisherbrand™ FB67891
Description
- Class F2 stainless steel calibration weight
- Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
- In compliance with OIML R111
Specifications
F2 | |
1 kg | |
Cylindrical |
Stainless Steel | |
1 kg |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F2 > 1kg
Spot an opportunity for improvement?Share a Content Correction