Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F2
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
Brand:  Fisherbrand™ FB67895
 View more versions of this product
Product Code. 15545179
  • £202.00 / 1 unit
Estimated Shipment: 24-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class F2 stainless steel calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • In compliance with OIML R111
Specifications
Specifications
F2
2 kg
Cylindrical
Stainless Steel
2 kg