Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Brand: Fisherbrand™ FB67881
Description
- Class M1 stainless steel calibration weight
- In compliance with OIML R111
Specifications
M1 | |
100 g | |
Cylindrical |
Stainless Steel | |
100 g |
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1 > 100g
Spot an opportunity for improvement?Share a Content Correction