Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1 with certificate
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Brand:  Fisherbrand™ FB67898
 View more versions of this product
Product Code. 15575179
  • £204.00 / 1 unit
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifications
Specifications
M1
2 kg
Cylindrical
Stainless Steel
2 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1 with certificate > 2kg