Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1 with certificate
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
Brand:  Fisherbrand™ FB67910
 View more versions of this product
Product Code. 15595189
  • £760.00 / 1 unit
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifications
Specifications
M1
10 kg
Cylindrical
Stainless Steel
10 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1 with certificate > 10kg