Learn More
Fisherbrand™ Individual Stainless Steel Leaf Calibration Weights, Class E2 with certificate
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Brand: Fisherbrand™ FB67816
Description
- Class E2 calibration weight
- In compliance with OIML R111
- Delivered with a calibration certificate guaranteeing traceability
Spécification
E2 | |
Stainless Steel | |
20 mg | |
20 mg |