missing translation for 'onlineSavingsMsg'
Learn More

Indole-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

£18.80 - £165.00

Chemical Identifiers

CAS 487-89-8
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00005622
InChI Key OLNJUISKUQQNIM-UHFFFAOYSA-N
Synonym indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
PubChem CID 10256
ChEBI CHEBI:28238
IUPAC Name 1H-indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=CN2)C=O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10349343
View Documents
Thermo Scientific Acros
156640050
5 g Glass bottle
£18.80
5g
Please to purchase this item. Need a web account? Register with us today!
10553131
View Documents
Thermo Scientific Acros
156640250
25 g Plastic bottle
£57.40
25g
Please to purchase this item. Need a web account? Register with us today!
10512941
View Documents
Thermo Scientific Acros
156641000
100 g Plastic bottle
£165.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

487-89-8
145.16
OLNJUISKUQQNIM-UHFFFAOYSA-N
10256
1H-indole-3-carbaldehyde
C9H7NO
MFCD00005622
indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
CHEBI:28238
C1=CC=C2C(=C1)C(=CN2)C=O

Specifications

487-89-8
240°C
96% min. (HPLC)
MFCD00005622
indole-3-carboxaldehyde, 3-formylindole, 1h-indole-3-carboxaldehyde, indole-3-aldehyde, indole-3-carbaldehyde, indole-3-carboxyaldehyde, 3-indolecarboxaldehyde, beta-indolylaldehyde, 3-indolecarbaldehyde, indol-3-carboxaldehyde
OLNJUISKUQQNIM-UHFFFAOYSA-N
1H-indole-3-carbaldehyde
10256
145.16
Crystalline Powder
193°C to 199°C
Authentic
C9H7NO
21, 313
Solubility in water: insoluble
C1=CC=C2C(=C1)C(=CN2)C=O
145.16
CHEBI:28238
97%
Indole-3-carboxaldehyde, 97%
Videos
Safety and Handling

Safety and Handling

AIR SENSITIVE

missing translation for 'einecsNumber' : 207-665-8

missing translation for 'rtecsNumber' : NL5993600

missing translation for 'tsca' : TSCA

Product Identifier
  • Indole-3-carboxaldehyde

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only