missing translation for 'onlineSavingsMsg'
Learn More

Indole-4-methanol, 97%, Thermo Scientific™

£54.40 - £180.00

Chemical Identifiers

CAS 1074-85-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD01632220
InChI Key BVSGXWCTWBZFEV-UHFFFAOYSA-N
Synonym indole-4-methanol, 1h-indol-4-yl methanol, 1h-indole-4-methanol, 4-hydroxymethylindole, 4-hydroxymethyl-1h-indole, indol-4-ylmethan-1-ol, pubchem7973, 4-hydroxymethyl indole, 4-hydroxymethyl-indole, acmc-2098wi
PubChem CID 2773457
IUPAC Name 1H-indol-4-ylmethanol
SMILES OCC1=C2C=CNC2=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15325018
View Documents
Thermo Scientific Acros
455532500
250 mg
£54.40
250mg
Please to purchase this item. Need a web account? Register with us today!
15315018
View Documents
Thermo Scientific Acros
455530010
1 g
£180.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1074-85-7
147.18
BVSGXWCTWBZFEV-UHFFFAOYSA-N
2773457
OCC1=C2C=CNC2=CC=C1
C9H9NO
MFCD01632220
indole-4-methanol, 1h-indol-4-yl methanol, 1h-indole-4-methanol, 4-hydroxymethylindole, 4-hydroxymethyl-1h-indole, indol-4-ylmethan-1-ol, pubchem7973, 4-hydroxymethyl indole, 4-hydroxymethyl-indole, acmc-2098wi
1H-indol-4-ylmethanol

Specifications

1074-85-7
96% min. (GC)
MFCD01632220
BVSGXWCTWBZFEV-UHFFFAOYSA-N
1H-indol-4-ylmethanol
2773457
97%
46.0°C to 50.0°C
C9H9NO
indole-4-methanol, 1h-indol-4-yl methanol, 1h-indole-4-methanol, 4-hydroxymethylindole, 4-hydroxymethyl-1h-indole, indol-4-ylmethan-1-ol, pubchem7973, 4-hydroxymethyl indole, 4-hydroxymethyl-indole, acmc-2098wi
OCC1=C2C=CNC2=CC=C1
147.18
147.17
Indole-4-methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

Product Identifier
  • Indol-4-methanol

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only