missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indole-6-methanol, 97%, Thermo Scientific™
£64.32 - £173.87
Chemical Identifiers
CAS | 1075-26-9 |
---|---|
Molecular Formula | C9H9NO |
Molecular Weight (g/mol) | 147.18 |
MDL Number | MFCD02179595 |
InChI Key | WRMZOPANDOHWJU-UHFFFAOYSA-N |
Synonym | 6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol |
PubChem CID | 2773459 |
SMILES | OCC1=CC=C2C=CNC2=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15585896
|
Thermo Scientific Acros
455542500 |
250 mg |
£64.32
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15575896
|
Thermo Scientific Acros
455540010 |
1 g |
£173.87
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
1075-26-9 | |
147.18 | |
WRMZOPANDOHWJU-UHFFFAOYSA-N | |
2773459 |
C9H9NO | |
MFCD02179595 | |
6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol | |
OCC1=CC=C2C=CNC2=C1 |
Specifications
1075-26-9 | |
Authentic | |
C9H9NO | |
6-hydroxymethylindole, 1h-indol-6-yl methanol, indole-6-methanol, 1h-indole-6-methanol, indol-6-ylmethan-1-ol, indol-6-yl-methanol, pubchem7966, 6-hydroxymethyl indole, 6-indolmethanol | |
OCC1=CC=C2C=CNC2=C1 | |
147.18 | |
147.17 | |
Indole-6-methanol |
45.0°C to 50.0°C | |
96% min. (GC) | |
MFCD02179595 | |
WRMZOPANDOHWJU-UHFFFAOYSA-N | |
1H-indol-6-ylmethanol | |
2773459 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Indol-6-methanol
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Indole-6-methanol, 97%, Thermo Scientific™