missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Indomethacin

Product Code. 15472202 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15472202

Brand: Thermo Scientific Chemicals 458030250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Questo articolo non è restituibile. Consulta la politica di reso

Indomethacin, CAS # 53-86-1, is an indole-based nonsteroidal anti-inflammatory (NSAID). | CAS: 53-86-1 | C19H16ClNO4 | 357.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Indomethacin is a synthetic NSAID of the indole family that inhibits cyclooxygenase and reduces prostaglandin and pyrogen release

• Indomethacin may suppress the multi-drug resistant transporter and thereby increase the efficacy of antineoplastics

• Indomethacin activates phosphatases that inhibit signaling pathways associated with cancer cell migration and proliferation

Applications

• Indomethacin may be used for the laboratory study of cancer cell proliferation, tumorigenesis and drug interactions

• Indomethacin may be used for laboratory study of the effect of inhibiting the multi-drug resistance transporter, or for downregulation of survival signaling in cells via survivin

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 53-86-1
Molecular Formula C19H16ClNO4
Molecular Weight (g/mol) 357.79
InChI Key CGIGDMFJXJATDK-UHFFFAOYSA-N
Synonym indomethacin, indometacin, indocin, indometacine, indomethacine, indocid, metindol, amuno, indomethazine, imbrilon
PubChem CID 3715
ChEBI CHEBI:49662
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
SMILES CC1=C(C2=C(N1C(=O)C3=CC=C(C=C3)Cl)C=CC(=C2)OC)CC(=O)O

Specifica

Melting Point 155.0°C
Loss on Drying 0.5% max. (100°C, 2 h, 5 mm Hg)
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 357.79
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Indomethacin
compliance-icons compliance-icons
Identificativo prodotto
  • Indomethacin
Avvertenza
  • Danger
Categoria di pericolo
  • Acute toxicity Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
Indicazioni di pericolo
  • H300-Fatal if swallowed.
  • H317-May cause an allergic skin reaction.
  • H360FD-May damage fertility. May damage the unborn child.
Consigli di prudenza
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato