Learn More
Kartell™ Industrial Scoops
Designed for heavy industrial use
Brand:  Kartell™ 0111000
 View more versions of this product
Product Code. 15865497
  • £60.00 / Pack of 25
Estimated Shipment: 28-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made of HDPE (High-Density Polyethylene)
  • Features a round shaped design to facilitate complete transfer of sampling material
  • Suitable for foodstuff
Specifications
Specifications
HDPE
250 mL
25/Pk.
25 mm