missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Infectious Disease Epitope Microarray

Infectious Disease Epitope Microarray with 4344 epitope specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.037.001

Additional Details : Weight : 0.02200kg

Product Code. 16317778

  • £482.30 / Each
Estimated Shipment: 22-10-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: 3760 linear B-cell epitopes of 196 different pathogens referenced in the Immune Epitope Database and two selected Toxoplasma gondii antigens as overlapping peptides
  • Database Epitope Lengths: 4 to 44 aa
  • Peptide Lengths: 4 to 17 aa depending on the database epitope length; longer epitopes were translated into overlapping 17 aa peptides
  • Number of Peptides: 4344 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEPperSlide™ PEGMA/PMMA
Antibody validation, epitope mapping, serum screening, biomarker discovery
Microscopic Glass
Individual barcode for each microarray
1 Slide
1 mm
Transparent
75.4 x 25 mm
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers