missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Influenza Virus H1N1 Proteome Microarray

Influenza Virus H1N1 Proteome Microarray with 3555 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.015.001

Product Code. 16377758

  • £515.00 / Each
Estimated Shipment: 13-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Microarray Content: A: Hemagglutinin, B: Matrix protein 1, C: Matrix protein 2, D: Nucleoprotein, E: Nuclear export protein, F: Neuraminidase, G: Non-structural protein 1, H: Polymerase acidic protein, I: Protein PB1-F2, J: Polymerase basic protein 2, K: RNA-directed RNA polymerase catalytic subunit
  • Peptide Length/Overlap: 15 aa/ 12 aa
  • Number of Peptides: 3555 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEGMA/PMMA
1 Slide
1 mm
Transparent
Individual barcode for each microarray
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Microscopic Glass
Antibody validation, epitope mapping, serum screening, biomarker discovery
75.4 x 25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.