missing translation for 'onlineSavingsMsg'
Learn More

PEPperCHIP™ Influenza Virus H5N1 Proteome Microarray

Influenza Virus H5N1 Proteome Microarray with 5506 virus specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.

Brand:  PEPperCHIP™ SPC.016.001

Additional Details : Weight : 0.02200kg

Product Code. 16387758

  • £482.30 / Each
Estimated Shipment: 15-10-2024
to see stock.
Add to basket

Description

Description

  • Microarray Content: 11 selected antigens of 12 different strains of Influenza A virus (H5N1): A: Hemagglutinin, B: Matrix protein 1, C: Matrix protein 2, D: Nucleoprotein, E: Nuclear export protein, F: Neuraminidase, G: Non-structural protein 1, H: Polymerase acidic protein, I: Protein PB1-F2, J: Polymerase basic protein 2, K: RNA-directed RNA polymerase catalytic subunit
  • Peptide Length/Overlap: 15 aa/ 10 aa
  • Number of Peptides: 5506 printed in duplicate and HA, polio and c-Myc control peptides
Specifications

Specifications

PEPperSlide™ PEGMA/PMMA
Antibody validation, epitope mapping, serum screening, biomarker discovery
Microscopic Glass
Individual barcode for each microarray
1 Slide
1 mm
Transparent
75.4 x 25 mm
High-density Peptide Microarray
Non-sterile
1 array copy per microarray
Combinatorial peptide synthesis by amino acid laser printing
ISO 9001:2015
RUO
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers