missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Inhibitor Removal Resin

£23.32 - £258.00

Chemical Identifiers

CAS 9003-70-7
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396099
View Documents
Thermo Scientific Chemicals
042489.14
25 g
£23.32
25g
Estimated Shipment: 31-12-2024
to see stock.
Add to basket
 
11319018
View Documents
Thermo Scientific Chemicals
042489.22
100 g
£81.70
100g
Estimated Shipment: 20-12-2024
to see stock.
Add to basket
 
11309018
View Documents
Thermo Scientific Chemicals
042489.36
500 g
£258.00
500g
Estimated Shipment: 31-12-2024
to see stock.
Add to basket
 
Description

Description

Inhibitor removal resin is used for the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol. It is also take part in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9003-70-7

Specifications

Inhibitor Removal Resin
Odorless
9003-70-7
For the removal of phenolic inhibitors: hydroquinone, hydroquinone monomethyl ether, tert-butylcatechol
CC(C)(C)OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C=O
409.48
9003-70-7
C24H27NO5
Crystalline Solid
BCIPGSZQUDLGSY-UHFFFAOYNA-N
tert-butyl 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxopentanoate
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only