missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Injection Needle

With O.D. 0.7mm guidance

Brand:  Cytiva 18-1801-42

Product Code. 17342471

  • £167.00 / Pack of 12

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • With guidance of 0.7mm O.D.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Injection Needle
0.7 mm O.D. guidance
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers