Learn More
Thermo Scientific™ Injection Port Liners, Split/Splitless Mixed Liner Sample Pack
Use these injection port liners to optimize performance of Agilent GC instruments.
Brand: Thermo Scientific™ 453AG001
Code : 16
Additional Details : Weight : 0.10000kg
Description
Optimize the performance of Agilent™ GC systems with our specifically designed Thermo Scientific™ Injection Port Liners for Agilent instruments. Highly deactivated and produced to exacting tolerances, these injection liners ensure exceptional reproducibility.
- Highly deactivated borosilicate glass
- Quartz wool packing material
- ID × OD × L: 4 × 6.3 × 78.5mm
Specifications
453AG001 | |
Split/Splitless Mixed Liner Sample Pk. | |
4 | |
GC Injection Port |
Mixed | |
Split/Splitless Mixed Liner Sample Pack | |
Mixed |