Learn More
Miele Professional™ Injector Nozzle
Injector nozzles with plastic supports
Brand:  Miele Professional™ 3810400
41.56 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 10640384
  • £44.30 / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Suitable for standard mobile injector units and injector baskets
  • Suitable for measuring flasks and test tubes, for example
Specifications
Specifications
Injector Nozzle
Plastic support
1 Each
ID 240
6 dia. x 240 mm L