Learn More
B Braun™ Injekt Solo Cone Syringes
Enjoy the smooth, steady operation of the B Braun™ Solo Cone Leur syringe. Its green plunger enables easy recognition of filling volumes and its double-seal stopper keeps contents intact.
Brand:  B Braun™ 4606108V
 View more versions of this product
Product Code. 12742637
Description
Description
Enjoy the smooth, steady operation of the B Braun™ Solo Cone Leur syringe. Its green plunger enables easy recognition of filling volumes and its double-seal stopper keeps contents intact.
  • Injekt™
  • Transparent barrel with graduations
  • Green plunger
  • Barrel and plunger rod: Polypropylene
  • No DEHP or natural rubber
Specifications
Specifications
EN ISO 7886-1
Single-use
10 mL
Syringe
1/Ea.
Sterile