Learn More
METTLER TOLEDO™ InLab™ Expert pH Electrodes
A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.
Brand: METTLER TOLEDO™ 51343103
241.43 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Description
Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.
- For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
- Features a PEEK plastic shaft for harsh environments and requires no maintenance
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- General Lab
- Beverages
- Waste water
- Suspended solids
- Environmental
- TRIS compatible
Accessories & Care
- Open Junction and non-refillable - no additional electrolyte required
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Specifications
DIN 19262, 4 mm Banana Plug | |
PEEK | |
0°C to 100°C | |
Xerolyt Polymer | |
pH Meter | |
U Glass | |
PT1000 | |
FRISCOLYT-B | |
0°C to 100°C | |
47 in. | |
1.2 m | |
12 mm |
Dual | |
0 to 14 | |
Combination pH Electrode, Integrated Pt1000 Temperature Probe | |
Emulsions, suspensions, effluent water and solutions containing Proteins and Sulphides | |
ARGENTHAL | |
InLab™ Expert DIN | |
<20 sec. | |
0°F to 212°F | |
4.7 in. | |
120 mm | |
0.47 in. |
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Days
12Hours
18Minutes
52Seconds
39 -
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
42Hours
18Minutes
52Seconds
39
Your input is important to us. Please complete this form to provide feedback related to the content on this product.