Learn More
METTLER TOLEDO™ InLab™ Expert pH Electrodes
A combination pH electrode with PEEK plastic shaft for chemical and mechanical durability, and a fixed cable. Designed for use in a variety of applications.
£238.00 - £576.00
Specifications
Material PEEK
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Filling Solution Type Xerolyt Polymer
Internal Reference ARGENTHAL
View More Specs
Products 10
Product Code Brand Model Type Connector Types For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Model Type Connector Types For Use With (Equipment) Price Quantity & Availability  
10454353
View Documents Promotion Details
METTLER TOLEDO™
51343104
InLab™ Expert NTC30 Combination Electrode, Integrated Temperature Probe (NTC30) Multi-Pin Multi-Pin to BNC (30281896)
£279.76
Each
In Stock
to see stock.
Add to basket
10009552
View Documents Promotion Details
METTLER TOLEDO™
51343100
InLab™ Expert Combination pH Electrode S7 S7 to BNC (30281915)
£238.00
Each
In Stock
to see stock.
Add to basket
10610904
View Documents Promotion Details
METTLER TOLEDO™
51343103
InLab™ Expert DIN Combination pH Electrode, Integrated Pt1000 Temperature Probe DIN 19262, 4 mm Banana Plug pH Meter
£281.00
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10040073
missing translation for 'viewDocuments' Aktionsdetails
METTLER TOLEDO™
51343102
InLab™ Expert Pro-ISM (2 m) Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology BNC/RCA Fixed Cable to sensor
£431.00
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10454543
missing translation for 'viewDocuments' Aktionsdetails
METTLER TOLEDO™
51343105
InLab™ Expert Pt1000 Combination pH Electrode, Integrated Pt1000 Temperature Probe Multi-Pin Multi-Pin to BNC (30281896
£315.00
Each
Estimated Shipment: 14-04-2025
to see stock.
Zum Warenkorb hinzufügen
10660324
missing translation for 'viewDocuments' Aktionsdetails
METTLER TOLEDO™
51343101
InLab™ Expert Pro Combination Electrode, Integrated Temperature Probe BNC/RCA Fixed Cable to sensor
£358.00
Each
In Stock
to see stock.
Edge
Zum Warenkorb hinzufügen
10447144
missing translation for 'viewDocuments' Aktionsdetails
METTLER TOLEDO™
51340288
InLab™ Expert Go Combination Electrode, Integrated Temperature Probe BNC/RCA IP67 Fixed Cable to sensor
£392.00
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10431435
missing translation for 'viewDocuments' Aktionsdetails
METTLER TOLEDO™
51344102
InLab™ Expert Go-ISM Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology BNC/RCA IP67 Fixed Cable to sensor
£524.00
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
10594925
missing translation for 'viewDocuments' Aktionsdetails
METTLER TOLEDO™
51344104
InLab™ Expert Go-ISM (10 m) Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology BNC/RCA IP67 Fixed Cable to sensor
£576.00
Each
In Stock
to see stock.
Zum Warenkorb hinzufügen
11394203
missing translation for 'viewDocuments' Aktionsdetails
METTLER TOLEDO™
30014096
InLab™ Expert Pro-ISM Combination Electrode, Integrated Temperature Probe, Intelligent Sensor Management (ISM) Technology BNC/RCA Fixed Cable to sensor
£398.00
Each
In Stock
to see stock.
Edge
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Ideal for critical samples such as suspensions or samples of unknown composition. Mettler Toledo Expert Series pH and pH/ATC Combination Electrodes with open junction puts the reference electrode in direct contact with the measured medium, eliminating the possibility of contamination or blockage.
  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Features a PEEK plastic shaft for harsh environments and requires no maintenance
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
  • General Lab
  • Beverages
  • Waste water
  • Suspended solids
  • Environmental
  • TRIS compatible
Accessories & Care
  • Open Junction and non-refillable - no additional electrolyte required
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Spezifikation
Spezifikation
PEEK
0°C to 100°C
ARGENTHAL
120 mm
12 mm
0 to 14
Xerolyt Polymer
U Glass
0.47 in.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
METTLER TOLEDO™ InLab™ Expert pH Electrodes
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt