missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Pure pH Electrodes
A glass pH electrode designed for ion-weak-samples such as ultrapure water and deionized water.
£347.00 - £596.00
Specifications
Junction | Immovable Glass Sleeve |
---|---|
Material | Glass |
pH Range | 1 to 11 |
For Use With (Application) | Measuring samples at low temperatures or with low ion concentrations |
Internal Reference | ARGENTHAL with Ag+ trap |
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Type | Connector Types | For Use With (Equipment) | Price | Quantity & Availability | ||
11384213
|
METTLER TOLEDO™
51344172 |
InLab™ Pure Pro-ISM | Combination Electrode; Integrated Temperature Probe; Intelligent Sensor Management (ISM) Technology | Multi-Pin | Multi-Pin to BNC/RCA (30281896) |
£596.00
Each |
In Stock
Log in to see stock. |
||
15559601
|
METTLER TOLEDO™
30248112 |
InLab™ Pure | Combination Electrode | S7 | S7 to BNC (30281915) |
£347.00
Each |
In Stock
Log in to see stock. |
||
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Pure water
- Ultrapure water
- Deionized water
- Waste water
- Highly saline solution, sea water
- Low-ion strength
Accessories & Care
- Immovable glass sleeve junction
- LoT membrane glass
- InLab Pure: refillable FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
- InLab Pure Pro-ISM: refillable 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Specifications
Immovable Glass Sleeve | |
1 to 11 | |
ARGENTHAL with Ag+ trap | |
0°C to 80°C |
Glass | |
Measuring samples at low temperatures or with low ion concentrations | |
LoT Glass | |
12 mm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title