missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Reach pH Electrodes

A pH electrode with an integrated temperature sensor featuring an extended shaft for measuring a wide range of samples in large vessels.

Brand:  METTLER TOLEDO™ 51343061

 View more versions of this product

Product Code. 10049392

  • £423.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Pilot reactor
  • Tank and Barrel
  • Large samples
  • Deep vessels

Accessories & Care

  • Ceramic junction
  • HA glass membrane
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

pH electrode InLab Reach Pro-425
Ceramic
0 to 14
Combination Electrode; Integrated Temperature Probe (NTC 30 kΩ)
Accurate measurement in a wide range of sample types
ARGENTHAL with Ag+ trap
InLab™ Reach Pro-425
<20 sec.
32°F to 212°F
15.7 in.
0.47 in.
Multi-Pin
Glass
0°C to 100°C
3mol/L KCl
Multi-Pin to BNC (30281896)
HA Glass
NTC 30 kilohm
3mol/L KCl
0°C to 100°C
400 mm
12 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.