missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Routine pH Electrodes

Product Code. 15529611 Shop All Mettler Toledo Products
Click to view available options
Connector Types:
BNC IP67
Multi-Pin
S7
Includes:
Electrode only
ISM Temperature Probe
Temperature Probe
Temperature Probe (Pt1000)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15529611

Brand: METTLER TOLEDO™ 30248832

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A combination pH electrode with an integrated temperature sensor and chemically resistant HA glass body. It is designed for general lab applications and ideal for use at high pH values (IP67 rated).

  • For use with METTLER TOLEDO SevenExcellence, SevenCompact and portable meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Drinking water
  • Highly saline solutions or seawater
  • Microbiological samples
  • Corrosive acids and bases
  • General Lab
  • Chemical
  • High pH values
  • TRIS compatible

Accessories & Care

  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Tip: Open fill port while measuring, and also to refill electrolyte solution
  • Ceramic junction
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)

Order Info

Cable not included (sold separately).

Recommended Storage

InLab storage solution

TRUSTED_SUSTAINABILITY

Specifications

Description pH Electrode InLab Routine Go-ISM
Connector Types BNC IP67
Junction Ceramic
Material Glass
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
For Use With (Application) Mobile applications
For Use With (Equipment) Fixed Cable to sensor
Includes ISM Temperature Probe
Internal Reference 3 mol/L KCl
Membrane HA Glass
Model InLab™ Routine Go-ISM
Storage Requirements InLab Storage Solution
Temperature (Metric) Range 0°C to 100°C
Length (Metric) Cable 1.8 m
Length (Metric) Stem 120 mm
Diameter (Metric) Body 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.