missing translation for 'onlineSavingsMsg'
Learn More

METTLER TOLEDO™ InLab™ Science pH Electrodes

Product Code. p-3639391 Shop All Mettler Toledo Products
Click to view available options
Connector Types:
Multi-Pin
S7
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10522225

Brand: METTLER TOLEDO™ 51343070

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

A pH electrode featuring a special glass body which is resistant to corrosive or organic samples, and is ideal for high temperatures and biological media. The high electrolyte outflow prevents contaminants from entering and clogging the junction, which would interfere with the reading.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Waste water
  • TRIS buffer
  • Micro-biological samples
  • Yeast fermentation solutions
  • Starch solutions
  • Organic solvents
  • General lab

Accessories & Care

  • Moveable sleeve junction
  • Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types S7
Junction Moveable Glass Sleeve
Material Glass
pH Range 0 to 12
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type 3mol/L KCl
For Use With (Application) Slurries and emulsions
For Use With (Equipment) S7 to BNC (30281915)
Internal Reference ARGENTHAL with Ag+ trap
Membrane A41 Glass
Model InLab™ Science
Response Time <20 sec.
Storage Requirements 3mol/L KCl
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 4.7 in.
Length (Metric) 120 mm
Diameter (English) 0.47 in.
Diameter (Metric) 12 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.