missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METTLER TOLEDO™ InLab™ Surface pH Electrodes
Shop All Mettler Toledo Products
Click to view available options
Connector Types:
Multi-Pin
S7
Unit Size:
Each
Description
- For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. Connection cables may be required.
- Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
- Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
- Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.
Featured Applications
- Agar plates
- Drop size samples
- Paper
- Skin
- Leather
- Suspended solids
- Textiles and Prints
Accessories & Care
- Ceramic ring junction
- Polysulfone membrane
- Refillable 3M KCl (51350072): Keep probe filled with electrolyte for accurate results
- Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
- Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
Recommended Storage
InLab storage solution
Specifications
Specifications
| Connector Types | Multi-Pin |
| Junction | Ceramic |
| Material | Glass Shaft |
| pH Range | 1 to 11 |
| Type | Combination Electrode |
| For Use With (Application) | Measurement of solid or semi-solid samples |
| For Use With (Equipment) | Multi-Pin to BNC/RCA (30281896) |
| Includes | ISM Temperature Probe |
| Internal Reference | 3mol/L KCl |
| Membrane | LoT Glass |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction