missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Inlet Tubes for Autoclavable Bottletop Dispenser

For use with Fisherbrand Autoclavable Bottletop Dispenser. Inlet tube is used to transfer reagent from bottle to the dispenser. This tube is telescopic in nature, thus can be adapted for different sized bottles.

£15.90

Specifications

Certifications/Compliance CE Certified, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655
Color Translucent
For Use With (Equipment) Fisherbrand Autoclavable Bottletop Dispenser
View More Specs

Products 1
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
15867702
View Documents Promotion Details
Fisherbrand™
190 to 370 mm
£15.90
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
 
Description

Description

  • Made of FEP material
  • Telescopic tube; length is adjustable
  • Broader side: Outer dia. is 9mm, Inner dia. is 8mm
  • Narrower side: Outer dia. is 7mm, Inner dia. is 6mm
Storage: Store the instrument and accessories only in clean conditions in a cool and dry place. Storage temperature: from -20°C to 50°C (from -4°F to 122°F).Note: Tube has a small end and a large end. The smaller end can be used with 2.5ml, 5ml and 10ml Dispensers. The larger end is good for 30ml, 60ml and 100ml Dispensers.

Certifications

CE Certified, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655

Specifications

Specifications

CE Certified, ISO 13485-2003, ISO 9001-2008, Compliance as per ISO 8655
Translucent
Fisherbrand Autoclavable Bottletop Dispenser
Videos
SDS
Documents

Documents

Product Certifications