missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BINDER™ Inner Chamber, Reinforced
Reinforced inner chamber for Series FP Avantgarde.Line Drying and Heating Chambers. Choose from four types of maximum total load options: 85 kg, 170 kg, 300 kg, and 350 kg.
£1855.00
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
17874451
|
BINDER™
8012-2224 |
170 kg |
£1855.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
17884451
|
BINDER™
8012-2225 |
300 kg |
£1855.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
17894451
|
BINDER™
8012-2226 |
350 kg |
£1855.00
Each |
Estimated Shipment: 04-09-2024
Log in to see stock. |
|||||
Description
- Accessory for Series FP Avantgarde.Line Drying and Heating Chambers
Specifications
Inner Chamber | |
230 V |