Learn More
Kartell™ Inoculation Loops and Needles
Manufactured with high impact polystyrene for good dimensional stability. Kartell™ Inoculation Loops and Needles provide a two–in–one sterile disposable tool for inoculation.
Brand:  Kartell™ 0265100
Product Code. 10040941
  • ONLINE EXCLUSIVE PRICE
    £529.00
    £519.70 / Pack of 8000
    Save £9.30
    1% Off
Only null left
Estimated Shipment: 23-02-2026
Add to basket
Description
Description
  • Gamma sterilized and packed in polybags
  • Looped end to retrieve an inoculum from a culture, transfer inoculum for streaking or transfer microscopic organisms
  • Needle end for picking off individual isolated colonies  or making stab inoculations into agar slants or tubes of solid culture medium
  • CE–IVD