missing translation for 'onlineSavingsMsg'
Learn More

WTW™ inoLab™ Multi 9620 IDS Multi-parameter Benchtop Meter Set C
GREENER_CHOICE

Use this Benchtop meter for simultaneous measurement with two independent measuring channels. WTW™ inoLab™ Multi 9620 IDS Multi-parameter Benchtop Meter Set C is wireless ready and offers fatigue proof reading on brilliant color display.

Brand:  WTW™ 1FD56C

2607.75 GBP valid until 2024-12-15
Use promo code "21658" to get your promotional price.


Product Code. 15562016

  • £2775.00 / Each
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Digital high performance multi-parameter device with two inputs for pH, ISE, conductivity and dissolved oxygen and comprehensive GLP functions
  • Includes DS sensors: digital IDS pH electrode SenTix™ 980, buffer 4, 7 and 10.01, 3 mol/L KCl, digital IDS conductivity cell TetraCon™ 925, 0.01 mol/L KCl, conductivity standard
  • Modern high-performance multi-parameter meter with two galvanically isolated measuring channels pH/mV, ISE (including incremental methods), conductivity and oxygen measurement
  • With a large 16:9 color graphic display under a hygienic glass shield and antibacterial keyboard for use, even in critical areas
  • Use the new wireless modules together with the new IDS plug head sensors, be independent from cables and measure i.e. conveniently under laboratory hoods or laminar flow benches
  • The massive zinc diecasting lower housing adds safety against interference and ensure a safe, stable stand on the lab bench
  • Two USB interfaces to communicate with the computer, printer or USB memory stick allow the transfer of all data
  • An activatizble user administration expands the automatic GLP documentation by additional safety and traceability
Integratable adapter available for conventional pH/ORP/ISE electrodes. The scope of delivery includes the MultiLab™ Importer to capture data via Excel™ as well as the MultiLab™ User (user administration).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

1
1 to 5
LCD Color Graphic
Universal power supply 100 to 240V, 50/60Hz
All except ORP
2 to 7 (requires adapter)
2
Manual 500/10000 automatic
Multi-parameter Benchtop Meter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers