missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Inorganic Pyrophosphatase/PPA1 Protein

Highly purified. Generating reliable and reproducible results.

£829.00

Specifications

For Use With (Application) SDS-PAGE
Formulation Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 50mM NaCl
Molecular Weight (g/mol) 21.9kDa
Purification Method SDS-PAGE
Quantity 0.5 mg
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18573342
View Documents
Novus Biologicals™
NBP1-72378-0.5mg
0.5 mg
£829.00
0.50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-176 of E.coli Inorganic Pyrophosphatase/PPA1 The Recombinant E. coli Inorganic Pyrophosphatase/PPA1 Protein is derived from E. coli. The Recombinant E. coli Inorganic Pyrophosphatase/PPA1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

SDS-PAGE
21.9kDa
0.5 mg
Store at -80°C. Avoid freeze-thaw cycles.
Inorganic Pyrophosphatase/PPA1
>95%
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 50mM NaCl
SDS-PAGE
Bacteria
RUO
Unconjugated
Videos
SDS
Documents

Documents

Product Certifications