missing translation for 'onlineSavingsMsg'
Learn More

Inosine 5'-monophophate disodium salt, ≥98%, MP Biomedicals™

Product Code. 11328889
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4691-65-0
Molecular Formula C10H11N4Na2O8P
Molecular Weight (g/mol) 392.17
MDL Number MFCD00036201,MFCD00036201
InChI Key AANLCWYVVNBGEE-IDIVVRGQSA-L
Synonym disodium 5'-inosinate
PubChem CID 123134061
IUPAC Name disodium [(2R,3S,4R,5R)-3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methyl phosphate
SMILES [Na+].[Na+].O[C@@H]1[C@@H](COP([O-])([O-])=O)O[C@H]([C@@H]1O)N1C=NC2=C1NC=NC2=O

Specifications

Color White
Quantity 5 g
Formula Weight 392.2
Physical Form Crystalline Powder
Chemical Name or Material Inosine 5'-monophophate disodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'