missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Inositol, 98+%
£38.00 - £708.00
Chemical Identifiers
CAS | 87-89-8 |
---|---|
Molecular Formula | C6H12O6 |
Molecular Weight (g/mol) | 180.16 |
MDL Number | MFCD00077932 |
InChI Key | CDAISMWEOUEBRE-UHFFFAOYSA-N |
Synonym | scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol |
PubChem CID | 892 |
ChEBI | CHEBI:24848 |
IUPAC Name | cyclohexane-1,2,3,4,5,6-hexol |
SMILES | C1(C(C(C(C(C1O)O)O)O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10784621
|
Thermo Scientific Chemicals
122261000 |
100 g | Plastic bottle |
£38.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10172630
|
Thermo Scientific Chemicals
122265000 |
500 g | Plastic bottle |
£115.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10297583
|
Thermo Scientific Chemicals
122260010 |
1 kg | Plastic bottle |
£201.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10338843
|
Thermo Scientific Chemicals
122260051 |
5 kg | Plastic drum |
£708.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
87-89-8 | |
180.16 | |
CDAISMWEOUEBRE-UHFFFAOYSA-N | |
892 | |
cyclohexane-1,2,3,4,5,6-hexol |
C6H12O6 | |
MFCD00077932 | |
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol | |
CHEBI:24848 | |
C1(C(C(C(C(C1O)O)O)O)O)O |
Specifications
87-89-8 | |
White | |
Authentic | |
C6H12O6 | |
06, II, 1157 | |
scyllo-inositol, myo-inositol, inositol, muco-inositol, epi-inositol, i-inositol, meso-inositol, allo-inositol, 1d-chiro-inositol, myoinositol | |
CDAISMWEOUEBRE-UHFFFAOYSA-N | |
cyclohexane-1,2,3,4,5,6-hexol | |
892 | |
180.16 | |
Crystalline Powder |
220°C to 228°C | |
0.5% max. | |
98% min. (ELSD) (HPLC) | |
MFCD00077932 | |
15, 5021 | |
(5% in water) Clear colorless | |
C1(C(C(C(C(C1O)O)O)O)O)O | |
180.16 | |
CHEBI:24848 | |
98+% | |
Inositol, 98+% |
Safety and Handling
missing translation for 'einecsNumber' : 201-781-2
missing translation for 'rtecsNumber' : NM7520800
missing translation for 'tsca' : TSCA
RUO – Research Use Only