Learn More
Greiner Bio-One Insect (Drosophila) Breeding Conical Container
Ideally suited for the cultivation of Drosophila melanogaster.
£111.00 - £231.00
Specifications
Bottom Shape Conical
Material Polystyrene
Type Drosophila Breeding Container
For Use With (Application) Cultivation of Drosophila melanogaster
View More Specs
Products 3
Product Code Brand Material Capacity (Metric) Price Quantity & Availability  
Product Code Brand Material Capacity (Metric) Price Quantity & Availability  
10517381
View Documents
Greiner Bio-One
199101
Polystyrene 16 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
13067683
View Documents
Greiner Bio-One
205101
Polystyrene 28 mL
£231.00
Pack of 1500
10160331
View Documents
Greiner Bio-One
217101
Polystyrene 68 mL
£113.00
Pack of 605
Description
Description
  • Bottom part of container made of either polystyrene or polypropylene
  • Compatible with gas-permeable ceaprene stopper made of water-repellent material
Specifications
Specifications
Conical
Polystyrene
Drosophila Breeding Container
Cultivation of Drosophila melanogaster