Learn More
Elma™ Insert Covers
Product Code. p-200004996
Click to view available options
Model:
100/120
150/180/300
180
30/40
60
80
Unit Size:
Each
Product Code. 17881357
Brand: Elma™ 1110262
Only null left
Add to basket
Add to basket
Insert cover contains two rubber rings and is designed for hanging cleaning cups or glasses in the cleaning basket.
  • Elma ultrasonic units can be operated with or without a cover
  • For more efficient operation, Elma offers various insert covers made of robust and easy-to-clean stainless steel
  • Covers make it possible to clean objects made of different materials simultaneously – but separately
  • If used correctly, all Elma inserts will help to prevent damage to the ultrasonic tank and thus extend the service life of the ultrasonic cleaner
  • Allows cleaning with aggressive fluids
  • Prolongs the service life of the unit
Specifications
Type Lid
For Use With (Equipment) Fisherbrand Select, Fisherbrand EASY, Elmasonic Med, Elmasonic P, Elmasonic S, Elmasonic Select, Elmasonic EASY
Material Stainless Steel
Model 30/40