missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Insert For Maxi Boxes

Manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes; these serve as an ideal solution for short-term storage of radioactive waste or radioisotopes.

Brand:  Fisherbrand™ CSR-R3F

 View more versions of this product

Product Code. 12347167

  • £39.90 / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Maxi Box Insert - 3 x Falcon tubes, 8 x 1.5ml tubes
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts that hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also supplied is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids
  • These serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, while the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
 
Specifications

Specifications

For Maxi Boxes
Insert
160mm
40mm
8 x 1.5mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.