missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INSL6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA554388
This item is not returnable.
View return policy
Description
Immunogen sequence: SARKLCGRYL VKEIEKLCGH ANWSQFRFEE ETPFSRLIAQ ASEKVEAYSP YQFESPQTAS PARGRGTNPV STSWEEAVNS WEMQSLPEYK D Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 42%.
The protein encoded by this gene contains a classical signature of the insulin superfamily and is significantly similar to relaxin and relaxin-like factor. This gene is preferentially expressed in testis. Its expression in testis is restricted to interstitial cells surrounding seminiferous tubules, which suggests a role in sperm development and fertilization.Specifications
INSL6 | |
Polyclonal | |
Unconjugated | |
INSL6 | |
INSL6; insulin like 6; insulin-like 6; insulin-like peptide 5; insulin-like peptide 6; Insulin-like peptide INSL6; Insulin-like peptide INSL6 A chain; Insulin-like peptide INSL6 B chain; Relaxin/insulin-like factor 1; RIF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11172 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y581 | |
INSL6 | |
Recombinant protein corresponding to Human INSL6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only