Learn More
Abeba™ Insoles 3551
Replaceable insole for shoes with air cushion sole is ESD dissipative and antistatic.
Brand:  Abeba™ 3551/47
Additional Details : Weight : 0.02500kg
 View more versions of this product
Product Code. 15620128
  • £11.77 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • Gray
  • Antistatic
  • ESD dissipative according to DIN EN 61340
  • Replaceable insole for shoes with air cushion sole
Specifications
Specifications
Gray
47
3551