Learn More
Abeba™ Insoles 3551
Replaceable insole for shoes with air cushion sole is ESD dissipative and antistatic.
Brand:  Abeba™ 3551/42
 View more versions of this product
Product Code. 15670118
  • £12.80 / 1 pair
Estimated Shipment: 12-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Gray
  • Antistatic
  • ESD dissipative according to DIN EN 61340
  • Replaceable insole for shoes with air cushion sole
Specifications
Specifications
Gray
42
3551