Learn More
Abeba™ Insoles 3565
Massage knobs stimulate circulation. ESD dissipative and antistatic.
Brand: Abeba™ 3565/38
Additional Details : Weight : 0.12000kg
Description
- Microfiber
- Anthracite
- ESD dissipative according to DIN EN 61340
- Antistatic
- Stimulating circulation by massage knobs, comfortable orthopaedic footbed
- Reflexor massage active-footbed
Specifications
Anthracite | |
Microfiber | |
38 | |
3565 |