Learn More
Abeba™ Insoles 3566
Massage knobs stimulate circulation. ESD dissipative and antistatic.
Brand:  Abeba™ 3566/42
Additional Details : Weight : 0.12000kg
 View more versions of this product
Product Code. 15630208
  • £13.56 / 1 pair
Estimated Shipment: 08-08-2024
to see stock.
Add to basket
Description
Description
  • Microfiber
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Stimulating circulation by massage knobs, comfortable orthopaedic footbed
  • Reflexor massage active-footbed
Specifications
Specifications
Microfiber
42
3566