Learn More
Abeba™ Insoles 3567
Product Code. p-7147583
Click to view available options
Size:
36
37
38
39
40
41
42
Unit Size:
1 pair
Product Code. 15680208
Brand: Abeba™ 3567/36
Only null left
Add to basket
Add to basket
ESD dissipative and antistatic insole is flat, smooth and antistatic.
  • Microfiber
  • White
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Flat, smooth
  • Footbed Reflexor
Specifications
Color White
Material Microfiber
Size 36
Style 3567