Learn More
Abeba™ Insoles 3572
Product Code. p-7147589
Click to view available options
Size:
36
37
38
39
40
41
42
43
Unit Size:
1 pair
Product Code. 15600258
Brand: Abeba™ 3572/36
Only null left
Add to basket
Add to basket
Massage knobs stimulate circulation. ESD dissipative and antistatic.
  • Microfiber
  • Anthracite
  • ESD dissipative according to DIN EN 61340
  • Antistatic
  • Stimulating circulation by massage knobs, comfortable orthopaedic footbed
  • Reflexor massage active-footbed
Specifications
Color Anthracite
Material Microfiber
Size 36
Style 3572