Learn More
Micronova™ Inspection Mirror Protective Storage Cover
Polysorb™ cover to protect and store Inspection Mirror
Brand: Micronova™ ISTSC11-66
Additional Details : Weight : 0.45000kg
Description
- Use with the highly polished stainless steel mirror for inspecting hard to reach areas
- A drawstring polyester cover
Specifications
QDIST-66P Inspection Mirror | |
Waffle Weave | |
Polyester Knit |