Learn More
Invitrogen™ InstantOne ELISA Wash Buffer (10X)
Developed for use with the InstantOne ELISA Cell Signaling assays. The InstantOne ELISA Wash Buffer is supplied as a 10x concentrate and is used for washing the InstantOne ELISA cell signaling assay microplate. It is a simple mix of buffers, salts and mild detergent.
Brand: Invitrogen™ IOWB1
Code : NEW
Additional Details : Weight : 0.47500kg
Description
- If washing wells with a microplate washer, use 3x washes with a 10-second mixing cycle
- Included with every InstantOne ELISA sold. Offered as a stand-alone item for cases where experimental procedures and/or automation require much higher volumes of wash buffer
Specifications
InstantOne ELISA Cell Signaling assays | |
Liquid | |
Wash Buffer |
8 mL | |
ELISA | |
Wash Buffer |
For Research Use Only. Not for use in diagnostic procedures.