Learn More
Instrument Calibration Standard 4 for ICP and HGA elements, Fluka™
Certified Reference Material, 100 mg/L each of As, Tl, 50 mg/L each of Cd, Se, 30 mg/L Pb in HNO3 5%
Brand: Honeywell Fluka E2106-100ML
Additional Details : CAS Number : 7697-37-2 Weight : 0.10000kg
Description
Honeywell Fluka™ analytical standards offers a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. Our products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Instrument Calibration Standard 4 for ICP and HGA elements | |
7778-39-4 | |
ICP | |
ICP-OES and ICP-MS |
Plastic Bottle | |
100 mL | |
<2 |