Learn More
Instrument Check Standards for ICP/MS, Set of 5, SPEX CertiPrep™
Instrument Check Standards for ICP-MS, 125 mL, Set of 5, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.
Brand:  SPEX Certiprep CL-ICS-SET
Product Code. 15698245
  • £762.00 / 1 set
Explore more special offers
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
  • Instrument Check Standards for ICP-MS
  • Set of 5
  • 125 mL Volume
Certifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Specifications
Specifications
Instrument Check Standards, Set of 5
Triple-leached LDPE bottle
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Trace ICP/ICP-MS
Set of 5
5 x 125 mL
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Set contains one 100mL bottle each of:
CL-ICS-1
CLHG2-1AY
CL-ICS-3
CL-ICS-4
CL-ICS-5
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product intended for laboratory use only