Learn More
Hamilton™ Terasaki Dispenser
This multi-syringe dispenser accurately dispenses six samples simultaneously up to 50 times with the press of a lever.
Brand:  Hamilton™ 201930
Product Code. 10723963
  • £1090.00 / Each
Explore more special offers
Description
Description
  • Designed for rapid filling of 60 well HLA/Terasaki plates with 6.35mm well spacing
  • Uses removable needles
  • Available for a range of volumes
Specifications
Specifications
1/Ea.
Dispenser
Borosilicate Glass, PTFE
250 μL