missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Instrument Tray

Buerkle™ — Buerkle™ Instrument Trays — ideal for various uses in laboratories, hospitals, pharmacies, the food and beverage industry

Brand:  Buerkle™ 4230-2550

32.85 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


 View more versions of this product

Product Code. 10000501

  • £43.10 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Melamine
  • White Tray without lid
  • Autoclavable
  • Lid available as accessory
Specifications

Specifications

5500 mL
White
350 mm
Instrument Tray
For Use in Laboratories, Hospitals, Food Industries and Areas in Which Cleanly Item Storage is Needed
3.93 in.
9.84 in.
350 x 250 x 100 mm
100 mm
Melamine
250 mm
13.77 in.
Rectangular
Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.