missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Insulin Rat ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ ERINSX10
Additional Details : Weight : 2.50000kg
Description
Insulin is a peptide hormone secreted by beta cells of the pancreatic islets. It regulates carbohydrate, protein and lipid metabolism by enhancing membrane transport of glucose, amino acids, and certain ions. It also promotes glycogen storage, formation of triglycerides and synthesis of proteins and nucleic acids. Insulin is 51-amino acid polypeptide product produced from a precursor peptide, proinsulin. Proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds and one molecule of C-peptide. Insulin deficiency results in diabetes mellitus, one of the leading causes of morbidity and mortality in the general population. Insulin is also present in tumors of b-cell origin such as insulinoma.Specifications
P01322, P01323 | |
5 μIU/mL | |
ELISA | |
Colorimetric Microplate Reader | |
Ins1, Ins2 | |
<10% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr 20 min |
4.69 to 300 μIU/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
24505, 24506 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
10 x 96 Tests | |
Plasma, 50 μL|Serum, 50 μL|Supernatant, 100 μL | |
Insulin | |
4 hr 45 min |